missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Methionine synthase reductase Protein
A DNA sequence encoding the Homo sapiens (Human) Methionine synthase reductase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8231-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002445.2 | |
Methionine synthase reductase Protein | |
Research Use Only | |
MTRR | |
Recombinant Protein | |
Yeast | |
MGAASVRAGA RLVEVALCSF TVTCLEVMRR FLLLYATQQG QAKAIAEEIC EQAVVHGFSA DLHCISESDK YDLKTETAPL VVVVSTTGTG DPPDTARKFV KEIQNQTLPV DFFAHLRYGL LGLGDSEYTY FCNGGKIIDK RLQELGARHF YDTGHADDCV GLELVVEPWI AGLWPALRKH FRSSRGQEEI SGALPVASPA SSRTDLVKSE LLHIESQVEL LRFDDSGRKD SEVLKQNAVN SNQSNVVIED FESSLTRSVP PLSQASLNIP GLPPEYLQVH LQESLGQEES QVSVTSADPV FQVPISKAVQ LTTNDAIKTT LLVELDISNT DFSYQPGDAF SVICPNSDSE VQSLLQRLQL EDKREHCVLL KIKADTKKKG ATLPQHIPAG CSLQFIFTWC LEIRAIPKKA FLRALVDYTS DSAEKRRLQE LCSKQGAADY SRFVRDACAC LLDLLLAFPS CQPPLSLLLE HLPKLQPRPY SCASSSLFHP GKLHFVFNIV EFLSTATTEV LRKGVCTGWL ALLVASVLQP NIHASHEDSG KALAPKISIS PRTTNSFHLP DDPSIPIIMV GPGTGIAPFI GFLQHREKLQ EQHPDGNFGA MWLFFGCRHK DRDYLFRKEL RHFLKHGILT HLKVSFSRDA PVGEEEAPAK YVQDNIQLHG QQVARILLQE NGHIYVCGDA KNMAKDVHDA LVQIISKEVG VEKLEAMKTL ATLKEEKRYL QDIWS | |
Greater than 90% as determined by SDS-PAGE. |
4552 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |