missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Legumain / LGMN Protein

A DNA sequence encoding the Homo sapiens (Human) Legumain/ LGMN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6301-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002300

  • 22559.24 NOK / 1mg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001008530.1
Legumain / LGMN Protein
1 mg
Not determined.
Full Length of Mature Protein
Human
His
Tris-based buffer, 50% glycerol
5641
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Lgmn
Recombinant Protein
E. coli
VPIDDPEDGG KHWVVIVAGS NGWYNYRHQA DACHAYQIIH RNGIPDEQIV VMMYDDIAYS EDNPTPGIVI NRPNGTDVYQ GVPKDYTGED VTPQNFLAVL RGDAEAVKGI GSGKVLKSGP QDHVFIYFTD HGSTGILVFP NEDLHVKDLN ETIHYMYKHK MYRKMVFYIE ACESGSMMNH LPDNINVYAT TAANPRESSY ACYYDEKRST YLGDWYSVNW MEDSDVEDLT KETLHKQYHL VKSHTNTSHV MQYGNKTIST MKVMQFQGMK RKASSPVPLP PVTHLDLTPS PDVPLTIMKR KLMNTN
Videos
SDS
Documents
Certificates
Special Offers

Special Offers