missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KCNJ10 Protein

A DNA sequence encoding the Homo sapiens (Human) KCNJ10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6256-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011970

  • 5325.00 NOK / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002232.2
KCNJ10 Protein
Research Use Only
KCNJ10
Recombinant Protein
E. coli
FLAKIARPKK RAETIRFSQH AVVASHNGKP CLMIRVANMR KSLLIGCQVT GKLLQTHQTK EGENIRLNQV NVTFQVDTAS DSPFLILPLT FYHVVDETSP LKDLPLRSGE GDFELVLILS GTVESTSATC QVRTSYLPEE ILWGYEFTPA ISLSASGKYI ADFSLFDQVV KVASPSGLRD STVRYGDPEK LKLEESLREQ AEKEGSALSV RISNV
Greater than 90% as determined by SDS-PAGE.
3766
100 μg
Not determined.
Cytoplasmic Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers