Learn More
enQuireBio™ Recombinant Human IL-17AF Protein
Recombinant Protein
1125.00 NOK - 74865.00 NOK
Specifications
Accession Number | NP_002181.1 |
---|---|
Gene ID (Entrez) | 3605 |
Name | IL-17AF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995649
|
enQuireBio™
QP5200-5UG |
5 μg |
1125.00 NOK
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975649
|
enQuireBio™
QP5200-25UG |
25 μg |
2650.00 NOK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955649
|
enQuireBio™
QP5200-100UG |
100 μg |
9910.00 NOK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985649
|
enQuireBio™
QP5200-500UG |
500 μg |
45570.00 NOK
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965649
|
enQuireBio™
QP5200-1MG |
1 mg |
74865.00 NOK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_002181.1 | |
IL-17AF Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Production of IL-6 from mouse 3T3 cells was significantly induced by less than 50 ng/ml of Human IL-17AF, or activity of at least 2.0 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
3605 | |
Research Use Only | |
IL17 | |
Recombinant Protein | |
E. coli | |
IL-17A:MIV KAGITIPRNP GCPNSEDKNF PRTVMVNLNI HNRNTNTNPK RSSDYYNRST SPWNLHRNED PERYPSVIWE AKCRHLGCIN ADGNVDYHMN SVPIQQEILV LRREPPHCPN SFRLEKILVS VGCTCVTPIV HHVA IL-17F :MRKIPKVGH TFFQKPESCP PVPGGSMKLD IGIINENQRV SMSRNIESRS TSPWNYTVTW DPNRYPSEVV QAQCRNLGCI NAQGKEDISM NSVPIQQETL VVRRKHQGCS VSFQLEKVLV TVGCTCVTPV IHHVQ | |
Greater than 95% as determined by SDS-PAGE. |