missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Gamma-synuclein Protein

A DNA sequence encoding the Homo sapiens (Human) Gamma-synuclein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6707-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005270

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001317049.1
Gamma-synuclein Protein
Research Use Only
SNCG
Recombinant Protein
E. coli
MDVFKKGFSI AKEGVVGAVE KTKQGVTEAA EKTKEGVMYV GAKTKENVVQ SVTSVAEKTK EQANAVSEAV VSSVNTVATK TVEEAENIAV TSGVVRKEDL RPSAPQQEGE ASKEKEEVAE EAQSGGD
Greater than 90% as determined by SDS-PAGE.
6623
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers