missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GABARAP / Apg8p1 Protein

A DNA sequence encoding the Homo sapiens (Human) GABARAP / Apg8p1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8508-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005021

  • 19129.63 NOK / 1mg


Estimated Shipment
19-11-2020
Add to basket

Specifications

Specifications

NP_009209.1
GABARAP / Apg8p1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
11337
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GABARAP
Recombinant Protein
E. coli
KFVYKEEHPF EKRRSEGEKI RKKYPDRVPV IVEKAPKARI GDLDKKKYLV PSDLTVGQFY FLIRKRIHLR AEDALFFFVN NVIPPTSATM GQLYQEHHEE DFFLYIAYSD ESVYGL
Videos
Documents