missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Four and a half LIM domains protein 1

A DNA sequence encoding the Homo sapiens (Human) Four and a half LIM domains protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7560-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009390

  • 22559.24 NOK / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001230807.1
Four and a half LIM domains protein 1
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
2275
Greater than 80% as determined by SDS-PAGE.
Research Use Only
FHL3
Recombinant Protein
E. coli
SESFDCAKCN ESLYGRKYIQ TDSGPYCVPC YDNTFANTCA ECQQLIGHDS RELFYEDRHF HEGCFRCCRC QRSLADEPFT CQDSELLCND CYCSAFSSQC SACGETVMPG SRKLEYGGQT WHEHCFLCSG CEQPLGSRSF VPDKGAHYCV PCYENKFAPR CARCSKTLTQ GGVTYRDQPW HRECLVCTGC QTPLAGQQFT SRDEDPYCVA CFGELFAPKC SSCKRPIVGL GGGKYVSFED RHWHHNCFSC ARCSTSLVGQ GFVPDGDQVL CQGCSQAGP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers