missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroblast growth factor 19 Protein
A DNA sequence encoding the Homo sapiens (Human) Fibroblast growth factor 19, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8543-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005108.1 | |
Fibroblast growth factor 19 Protein | |
Research Use Only | |
FGF19 | |
Recombinant Protein | |
E. coli | |
PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK | |
Greater than 90% as determined by SDS-PAGE. |
9965 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |