missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dystonin Protein

A DNA sequence encoding the Homo sapiens (Human) Dystonin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7837-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011091

  • 4941.08 NOK / 100µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138241.1
Dystonin Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
667
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DST
Recombinant Protein
E. coli
MAGYLSPAAY LYVEEQEYLQ AYEDVLERYK DERDKVQKKT FTKWINQHLM KVRKHVNDLY EDLRDGHNLI SLLEVLSGDT LPREKGRMRF HRLQNVQIAL DYLKRRQVKL VNIRNDDITD GNPKLTLGLI WTIILHFQIS DIHVTGESED MSAKERLLLW TQQATEGYAG IRCENFTTCW RDGKLFNAII HKYRP
Videos
SDS
Documents
Certificates
Special Offers

Special Offers