missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Deoxyribonuclease gamma Protein

A DNA sequence encoding the Homo sapiens (Human) Deoxyribonuclease gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7580-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009540

  • 34365.00 NOK / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001243489.1
Deoxyribonuclease gamma Protein
Research Use Only
DNASE1L3
Recombinant Protein
E. coli
MRICSFNVRS FGESKQEDKN AMDVIVKVIK RCDIILVMEI KDSNNRICPI LMEKLNRNSR RGITYNYVIS SRLGRNTYKE QYAFLYKEKL VSVKRSYHYH DYQDGDADVF SREPFVVWFQ SPHTAVKDFV IIPLHTTPET SVKEIDELVE VYTDVKHRWK AENFIFMGDF NAGCSYVPKK AWKNIRLRTD PRFVWLIGDQ EDTTVKKSTN CAYDRIVLRG QEIVSSVVPK SNSVFDFQKA YKLTEEEALD VSDHFPVEFK LQSSRAFTNS KKSVTLRKKT KSKRS
Greater than 90% as determined by SDS-PAGE.
1776
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers