missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome b-c1 complex subunit 8 Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-c1 complex subunit 8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8458-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004741

  • 12152.88 NOK / 500µg


Estimated Shipment
13-11-2020
Add to basket

Specifications

Specifications

NP_055217.2
Cytochrome b-c1 complex subunit 8 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
27089
Greater than 90% as determined by SDS-PAGE.
Research Use Only
QP-C/UQCRQ
Recombinant Protein
E. coli
GREFGNLTRM RHVISYSLSP FEQRAYPHVF TKGIPNVLRR IRESFFRVVP QFVVFYLIYT WGTEEFERSK RKNPAAY
Videos
Documents