missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cysteine dioxygenase type 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cysteine dioxygenase type 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7589-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001310494.1 | |
Cysteine dioxygenase type 1 Protein | |
Research Use Only | |
CDO1 | |
Recombinant Protein | |
E. coli | |
MEQTEVLKPR TLADLIRILH QLFAGDEVNV EEVQAIMEAY ESDPTEWAMY AKFDQYRYTR NLVDQGNGKF NLMILCWGEG HGSSIHDHTN SHCFLKMLQG NLKETLFAWP DKKSNEMVKK SERVLRENQC AYINDSIGLH RVENISHTEP AVSLHLYSPP FDTCHAFDQR TGHKNKVTMT FHSKFGIRTP NATSGSLENN | |
Greater than 80% as determined by SDS-PAGE. |
1036 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |