missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNTF Protein
A DNA sequence encoding the Homo sapiens (Human) CNTF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8530-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000605.1 | |
CNTF Protein | |
Research Use Only | |
CNTF | |
Recombinant Protein | |
E. coli | |
TEHSPLTPHR RDLCSRSIWL ARKIRSDLTA LTESYVKHQG LNKNINLDSA DGMPVASTDQ WSELTEAERL QENLQAYRTF HVLLARLLED QQVHFTPTEG DFHQAIHTLL LQVAAFAYQI EELMILLEYK IPRNEADGMP INVGDGGLFE KKLWGLKVLQ ELSQWTVRSI HDLRFISSHQ TGIPARGSHY IAN | |
Greater than 90% as determined by SDS-PAGE. |
1270 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |