missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD106 / VCAM1 Protein
A DNA sequence encoding the Homo sapiens (Human) CD106 / VCAM1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6877-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001069.1 | |
CD106 / VCAM1 Protein | |
Research Use Only | |
VCAM1 | |
Recombinant Protein | |
E. coli | |
FKIETTPESR YLAQIGDSVS LTCSTTGCES PFFSWRTQID SPLNGKVTNE GTTSTLTMNP VSFGNEHSYL CTATCESRKL EKGIQVEIYS FPKDPEIHLS GPLEAGKPIT VKCSVADVYP FDRLEIDLLK GDHLMKSQEF LEDADRKSLE TKSLEVTFTP VIEDIGKVLV CRAKLHIDEM DSVPTVRQAV KELQVYISPK NTVISVNPST KLQEGGSVTM TCSSEGLPAP EIFWSKKLDN GNLQHLSGNA TLTLIAMRME DSGIYVCEGV NLIGKNRKEV ELIVQEKPFT VEISPGPRIA AQIGDSVMLT CSVMGCESPS FSWRTQIDSP LSGKVRSEGT NSTLTLSPVS FENEHSYLCT VTCGHKKLEK GIQVELYSFP RDPEIEMSGG LVNGSSVTVS CKVPSVYPLD RLEIELLKGE TILENIEFLE DTDMKSLENK SLEMTFIPTI EDTGKALVCQ AKLHIDDMEF EPKQRQSTQT LYVNVAPRDT TVLVSPSSIL EEGSSVNMTC LSQGFPAPKI LWSRQLPNGE LQPLSENATL TLISTKMEDS GVYLCEGINQ AGRSRKEVEL IIQVTPKDIK LTAFPSESVK EGDTVIISCT CGNVPETWII LKKKAETGDT VLKSIDGAYT IRKAQLKDAG VYECESKNKV GSQLRSLTLD VQGRENNKDY FSPE | |
Greater than 90% as determined by SDS-PAGE. |
7412 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |