missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCL-W / BCL2L2 Protein

A DNA sequence encoding the Homo sapiens (Human) BCL-W / BCL2L2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7839-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011111

  • 22559.24 NOK / 1mg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001186768.1
BCL-W / BCL2L2 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
599
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BCL2L2
Recombinant Protein
E. coli
ATPASAPDTR ALVADFVGYK LRQKGYVCGA GPGEGPAADP LHQAMRAAGD EFETRFRRTF SDLAAQLHVT PGSAQQRFTQ VSDELFQGGP NWGRLVAFFV FGAALCAESV NKEMEPLVGQ VQEWMVAYLE TQLADWIHSS GGWAEFTALY GDGALEEARR LREGNWASVR TVLTGAVALG ALVTVGAFFA SK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers