missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP-dependent RNA helicase DDX19A Protein

A DNA sequence encoding the Homo sapiens (Human) ATP-dependent RNA helicase DDX19A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8225-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16023391

  • 5325.00 NOK / 100µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001307451.1
ATP-dependent RNA helicase DDX19A Protein
Research Use Only
DDX19A
Recombinant Protein
E. coli
MATDSWALAV DEQEAAVKSM TNLQIKEEKV KADTNGIIKT STTAEKTDEE EKEDRAAQSL LNKLIRSNLV DNTNQVEVLQ RDPNSPLYSV KSFEELRLKP QLLQGVYAMG FNRPSKIQEN ALPMMLAEPP QNLIAQSQSG TGKTAAFVLA MLSRVEPSDR YPQCLCLSPT YELALQTGKV IEQMGKFYPE LKLAYAVRGN KLERGQKISE QIVIGTPGTV LDWCSKLKFI DPKKIKVFVL DEADVMIATQ GHQDQSIRIQ RMLPRNCQML LFSATFEDSV WKFAQKVVPD PNVIKLKREE ETLDTIKQYY VLCSSRDEKF QALCNLYGAI TIAQAMIFCH TRKTASWLAA ELSKEGHQVA LLSGEMMVEQ RAAVIERFRE GKEKVLVTTN VCARGIDVEQ VSVVINFDLP VDKDGNPDNE TYLHRIGRTG RFGKRGLAVN MVDSKHSMNI LNRIQEHFNK KIERLDTDDL DEIEKIAN
Greater than 90% as determined by SDS-PAGE.
55308
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers