missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATG14 Protein

A DNA sequence encoding the Homo sapiens (Human) ATG14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7740-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16020531

  • 22559.24 NOK / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055739.2
ATG14 Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
22863
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ATG14
Recombinant Protein
E. coli
MASPSGKGAR ALEAPGCGPR PLARDLVDSV DDAEGLYVAV ERCPLCNTTR RRLTCAKCVQ SGDFVYFDGR DRERFIDKKE RLSRLKSKQE EFQKEVLKAM EGKWITDQLR WKIMSCKMRI EQLKQTICKG NEEMEKNSEG LLKTKEKNQK LYSRAQRHQE KKEKIQRHNR KLGDLVEKKT IDLRSHYERL ANLRRSHILE LTSVIFPIEE VKTGVRDPAD VSSESDSAMT SSTVSKLAEA RRTTYLSGRW VCDDHNGDTS ISITGPWISL PNNGDYSAYY SWVEEKKTTQ GPDMEQSNPA YTISAALCYA TQLVNILSHI LDVNLPKKLC NSEFCGENLS KQKFTRAVKK LNANILYLCF SQHVNLDQLQ PLHTLRNLMY LVSPSSEHLG RSGPFEVRAD LEESMEFVDP GVAGESDESG DERVSDEETD LGTDWENLPS PRFCDIPSQS VEVSQSQSTQ ASPPIASSSA GGMISSAAAS VTSWFKAYTG HR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers