missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ameloblastin Protein
A DNA sequence encoding the Homo sapiens (Human) Ameloblastin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9909-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057603.1 | |
Ameloblastin Protein | |
Research Use Only | |
AMBN | |
Recombinant Protein | |
Yeast | |
VPFFPQQSGT PGMASLSLET MRQLGSLQRL NTLSQYSRYG FGKSFNSLWM HGLLPPHSSL PWMRPREHET QQYEYSLPVH PPPLPSQPSL KPQQPGLKPF LQSAAATTNQ ATALKEALQP PIHLGHLPLQ EGELPLVQQQ VAPSDKPPKP ELPGVDFADP QGPSLPGMDF PDPQGPSLPG LDFADPQGST IFQIARLISH GPMPQNKQSP LYPGMLYVPF GANQLNAPAR LGIMSSEEVA GGREDPMAYG AMFPGFGGMR PGFEGMPHNP AMGGDFTLEF DSPVAATKGP ENEEGGAQGS PMPEANPDNL ENPAFLTELE PAPHAGLLAL PKDDIPGLPR SPSGKMKGLP SVTPAAADPL MTPELADVYR TYDADMTTSV DFQEEATMDT TMAPNSLQTS MPGNKAQEPE MMHDAWHFQE P | |
Greater than 90% as determined by SDS-PAGE. |
258 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |