Learn More
Novus Biologicals Recombinant Human alpha-Synuclein Active, Monomer, (Type 1), A53T Mutant Protein
Brand:  Novus Biologicals NBP3-14766-100UG
 View more versions of this product
Product Code. 18329266
  • 4619.75 NOK / 100µg
Explore more special offers
Description
Description
100 uM A53T alpha-Synuclein protein monomer seeded with 10 uM A53T alpha-Synuclein protein pre-formed fibrils in 25 uM Thioflavin T (PBS pH 7.4, 100 ul reaction volume) generated a fluorescence intensity of 28 000 RFU after incubation at 37C with shaking at 600 rpm for 56 hours. Fluorescence measured by excitation at 450 nm and emission at 485 nm on a microplate reader.
Specifications
Specifications
In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
6622
Store at -80C. Avoid freeze-thaw cycles.
SNCA
>95%, by SDS-PAGE
PBS pH 7.4
100 μg
alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), truncated alpha synuclein
Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience
alpha-Synuclein