missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alkaline Phosphatase Protein
A DNA sequence encoding the Homo sapiens (Human) Alkaline Phosphatase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8736-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001622.2 | |
Alkaline Phosphatase Protein | |
Research Use Only | |
ALPI | |
Recombinant Protein | |
E. coli | |
ENPAFWNRQA AEALDAAKKL QPIQKVAKNL ILFLGDGLGV PTVTATRILK GQKNGKLGPE TPLAMDRFPY LALSKTYNVD RQVPDSAATA TAYLCGVKAN FQTIGLSAAA RFNQCNTTRG NEVISVMNRA KQAGKSVGVV TTTRVQHASP AGTYAHTVNR NWYSDADMPA SARQEGCQDI ATQLISNMDI DVILGGGRKY MFPMGTPDPE YPADASQNGI RLDGKNLVQE WLAKHQGAWY VWNRTELMQA SLDQSVTHLM GLFEPGDTKY EIHRDPTLDP SLMEMTEAAL RLLSRNPRGF YLFVEGGRID HGHHEGVAYQ ALTEAVMFDD AIERAGQLTS EEDTLTLVTA DHSHVFSFGG YTLRGSSIFG LAPSKAQDSK AYTSILYGNG PGYVFNSGVR PDVNESESGS PDYQQQAAVP LSSETHGGED VAVFARGPQA HLVHGVQEQS FVAHVMAFAA CLEPYTACDL APPAC | |
Greater than 80% as determined by SDS-PAGE. |
248 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |