missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKT-interacting Protein
A DNA sequence encoding the Homo sapiens (Human) AKT-interacting protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8053-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001012398.1 | |
AKT-interacting Protein | |
Research Use Only | |
AKTIP | |
Recombinant Protein | |
E. coli | |
MNPFWSMSTS SVRKRSEGEE KTLTGDVKTS PPRTAPKKQL PSIPKNALPI TKPTSPAPAA QSTNGTHASY GPFYLEYSLL AEFTLVVKQK LPGVYVQPSY RSALMWFGVI FIRHGLYQDG VFKFTVYIPD NYPDGDCPRL VFDIPVFHPL VDPTSGELDV KRAFAKWRRN HNHIWQVLMY ARRVFYKIDT ASPLNPEAAV LYEKDIQLFK SKVVDSVKVC TARLFDQPKI EDPYAISFSP WNPSVHDEAR EKMLTQKKPE EQHNKSVHVA GLSWVKPGSV QPFSKEEKTV AT | |
Greater than 90% as determined by SDS-PAGE. |
64400 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |