missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AASDHPPT Protein

A DNA sequence encoding the Homo sapiens (Human) AASDHPPT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8073-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002511

  • 4941.08 NOK / 100µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_056238.2
AASDHPPT Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
60496
Greater than 90% as determined by SDS-PAGE.
Research Use Only
AASDHPPT
Recombinant Protein
E. coli
MVFPAKRFCL VPSMEGVRWA FSCGTWLPSR AEWLLAVRSI QPEEKERIGQ FVFARDAKAA MAGRLMIRKL VAEKLNIPWN HIRLQRTAKG KPVLAKDSSN PYPNFNFNIS HQGDYAVLAA EPELQVGIDI MKTSFPGRGS IPEFFHIMKR KFTNKEWETI RSFKDEWTQL DMFYRNWALK ESFIKAIGVG LGFELQRLEF DLSPLNLDIG QVYKETRLFL DGEEEKEWAF EESKIDEHHF VAVALRKPDG SRHQDVPSQD DSKPTQRQFT ILNFNDLMSS AVPMTPEDPS FWDCFCFTEE IPIRNGTKS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers