missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L9 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6622-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000652.2 | |
60S ribosomal protein L9 Protein | |
Research Use Only | |
RPL9 | |
Recombinant Protein | |
E. coli | |
MKTILSNQTV DIPENVDITL KGRTVIVKGP RGTLRRDFNH INVELSLLGK KKKRLRVDKW WGNRKELATV RTICSHVQNM IKGVTLGFRY KMRSVYAHFP INVVIQENGS LVEIRNFLGE KYIRRVRMRP GVACSVSQAQ KDELILEGND IELVSNSAAL IQQATTVKNK DIRKFLDGIY VSEKGTVQQA DE | |
Greater than 80% as determined by SDS-PAGE. |
6133 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |