missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L3 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8406-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004441

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000958.1
60S ribosomal protein L3 Protein
Research Use Only
RPL3
Recombinant Protein
E. coli
SHRKFSAPRH GSLGFLPRKR SSRHRGKVKS FPKDDPSKPV HLTAFLGYKA GMTHIVREVD RPGSKVNKKE VVEAVTIVET PPMVVVGIVG YVETPRGLRT FKTVFAEHIS DECKRRFYKN WHKSKKKAFT KYCKKWQDED GKKQLEKDFS SMKKYCQVIR VIAHTQMRLL PLRQKKAHLM EIQVNGGTVA EKLDWARERL EQQVPVNQVF GQDEMIDVIG VTKGKGYKGV TSRWHTKKLP RKTHRG
Greater than 90% as determined by SDS-PAGE.
6122
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers