missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L15 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8408-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001240308.1 | |
60S ribosomal protein L15 Protein | |
Research Use Only | |
RPL15 | |
Recombinant Protein | |
E. coli | |
GAYKYIQELW RKKQSDVMRF LLRVRCWQYR QLSALHRAPR PTRPDKARRL GYKAKQGYVI YRIRVRRGGR KRPVPKGATY GKPVHHGVNQ LKFARSLQSV AEERAGRHCG ALRVLNSYWV GEDSTYKFFE VILIDPFHKA IRRNPDTQWI TKPVHKHREM RGLTSAGRKS RGLGKGHKFH HTIGGSRRAA WRRRNTLQLH RYR | |
Greater than 90% as determined by SDS-PAGE. |
6138 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |