missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S16 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S16, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8410-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004461

  • 5325.00 NOK / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001011.1
40S ribosomal protein S16 Protein
Research Use Only
RPS16
Recombinant Protein
E. coli
PSKGPLQSVQ VFGRKKTATA VAHCKRGNGL IKVNGRPLEM IEPRTLQYKL LEPVLLLGKE RFAGVDIRVR VKGGGHVAQI YAIRQSISKA LVAYYQKYVD EASKKEIKDI LIQYDRTLLV ADPRRCESKK FGGPGARARY Q
Greater than 90% as determined by SDS-PAGE.
6217
100 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers