missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 40S ribosomal protein S13 Protein

A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S13, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8411-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004471

  • 12152.88 NOK / 500µg


Estimated Shipment
20-11-2020
Add to basket

Specifications

Specifications

NP_001008.1
40S ribosomal protein S13 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6207
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RPS13
Recombinant Protein
E. coli
RMHAPGKGLS QSALPYRRSV PTWLKLTSDD VKEQIYKLAK KGLTPSQIGV ILRDSHGVAQ VRFVTGNKIL RILKSKGLAP DLPEDLYHLI KKAVAVRKHL ERNRKDKDAK FRLILIESRI HRLARYYKTK RVLPPNWKYE SSTASALVA
Videos
Documents