missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Elongation factor P Protein

A DNA sequence encoding the Escherichia coli (strain K12) Elongation factor P, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7316-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008160

  • 38609.65 NOK / 1mg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418571.1
Elongation factor P Protein
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
948661
Greater than 90% as determined by SDS-PAGE.
Research Use Only
efp
Recombinant Protein
Yeast
ATYYSNDFRA GLKIMLDGEP YAVEASEFVK PGKGQAFARV KLRRLLTGTR VEKTFKSTDS AEGADVVDMN LTYLYNDGEF WHFMNNETFE QLSADAKAIG DNAKWLLDQA ECIVTLWNGQ PISVTPPNFV ELEIVDTDPG LKGDTAGTGG KPATLSTGAV VKVPLFVQIG EVIKVDTRSG EYVSRVK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers