missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L32 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L32, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8630-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005671

  • 3192.75 NOK / 10µg


Estimated Shipment
24-11-2020
Add to basket

Specifications

Specifications

NP_415607.1
50S ribosomal protein L32 Protein
10 μg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
31659212
Greater than 90% as determined by SDS-PAGE.
Research Use Only
rpmF
Recombinant Protein
E. coli
AVQQNKPTRS KRGMRRSHDA LTAVTSLSVD KTSGEKHLRH HITADGYYRG RKVIAK
Videos
SDS
Documents