missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S6 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8612-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418621.5 | |
30S ribosomal protein S6 Protein | |
Research Use Only | |
rpsF | |
Recombinant Protein | |
E. coli | |
MRHYEIVFMV HPDQSEQVPG MIERYTAAIT GAEGKIHRLE DWGRRQLAYP INKLHKAHYV LMNVEAPQEV IDELETTFRF NDAVIRSMVM RTKHAVTEAS PMVKAKDERR ERRDDFANET ADDAEAGDSE E | |
Greater than 90% as determined by SDS-PAGE. |
948723 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |