missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S15 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S15, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8619-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417634.1 | |
30S ribosomal protein S15 Protein | |
Research Use Only | |
rpsO | |
Recombinant Protein | |
E. coli | |
SLSTEATAKI VSEFGRDAND TGSTEVQVAL LTAQINHLQG HFAEHKKDHH SRRGLLRMVS QRRKLLDYLK RKDVARYTQL IERLGLRR | |
Greater than 90% as determined by SDS-PAGE. |
947686 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |