missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Serum amyloid A Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Serum amyloid A protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6646-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003050.1 | |
Serum amyloid A Protein | |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
E. coli | |
QWYSFVSEAA QGAWDMWRAY SDMREANYKN SDKYFHARGN YDAAQRGPGG AWAAKVISDA RENSQRITDL LRFGDSGHGA EDSKADQAAN EWGRSGKDPN HFRPAGLPDK Y | |
Greater than 90% as determined by SDS-PAGE. |
403585 | |
500 μg | |
Not determined. | |
Full Length | |
Dog | |
His-GST | |
Tris-based buffer, 50% glycerol |