missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 1 NS1 Protein
Recombinant Protein
4055.00 NOK - 14240.00 NOK
Specifications
Name | Serotype 1 NS1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue virus (Strain: Nauru/Western Pacific/1974) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943988
|
enQuireBio™
QP10475-100UG |
100 μg |
4055.00 NOK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953988
|
enQuireBio™
QP10475-500UG |
500 μg |
14240.00 NOK
500µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The NS1, non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.Specifications
Serotype 1 NS1 Protein | |
Untested | |
Dengue virus (Strain: Nauru/Western Pacific/1974) | |
Native | |
>95% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4. |