missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-3 Protein
A cDNA sequence encoding the Dengue Envelope-3 was constructed and used to recombinantly synthesize the protein.
1380.00 NOK - 73335.00 NOK
Specifications
Name | Dengue Envelope-3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15967858
|
enQuireBio™
QP11631-2UG |
2 μg |
1380.00 NOK
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947858
|
enQuireBio™
QP11631-10UG |
10 μg |
2140.00 NOK
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957858
|
enQuireBio™
QP11631-1MG |
1 mg |
73335.00 NOK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Especificaciones
Dengue Envelope-3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >90% pure as determined by 12.5% SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue Envelope-3 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |