missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus 65 kDa phosphoProtein

A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) 65 kDa phosphoprotein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7365-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16008470

  • 9233.45 NOK / 100µg
Estimated Shipment: 20-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

65 kDa phosphoProtein
100 μg
Not determined.
Partial (see sequence information for more details).
Cytomegalovirus
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
UL83
Recombinant Protein
E. coli
FDIDLLLQRG PQYSEHPTFT SQYRIQGKLE YRHTWDRHDE GAAQGDDDVW TSGSDSDEEL VTTERKTPRV TGGGAMAGAS TSAGRKRKSA SSATACTSGV MTRGRLKAES TVAPEEDTDE DSDNEIHNPA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers