missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cat Serum amyloid A Protein
A DNA sequence encoding the Felis catus (Cat) (Felis silvestris catus) Serum amyloid A protein, was expressed in the hosts and tags indicated.
3192.75 NOK - 34365.00 NOK
Specifications
Name | Cat Serum amyloid A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | SAA1 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084770
|
enQuireBio™
QP6645-EC-100UG |
100 μg |
9910.00 NOK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004780
|
enQuireBio™
QP6645-EC-1MG |
1 mg |
34365.00 NOK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094770
|
enQuireBio™
QP6645-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014780
|
enQuireBio™
QP6645-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024780
|
enQuireBio™
QP6645-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034780
|
enQuireBio™
QP6645-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Cat Serum amyloid A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-B2M | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
SAA1 | |
Recombinant Protein | |
E. coli | |
EWYSFLGEAA QGAWDMWRAY SDMREANYIG ADKYFHARGN YDAAQRGPGG AWAAKVISDA RENSQRVTDF FRHGNSGHGA EDSKADQEWG | |
Greater than 90% as determined by SDS-PAGE. |