Learn More
enQuireBio™ Recombinant Bovine Metalloproteinase inhibitor 4 Protein
A DNA sequence encoding the Bos taurus (Bovine) Metalloproteinase inhibitor 4, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6796-ye-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16015920
  • 11060.00 NOK / 100µg
Estimated Shipment: 18-06-2024
Specifications
NP_001039336.1
Metalloproteinase inhibitor 4 Protein
Research Use Only
TIMP4
Recombinant Protein
Yeast
CSCAPAHPQQ HVCHSALAIR AKISSEKVVP ASTDPADPQK MIRYEIKQIK MFKGFEKVND IQYIYTPFDS SLCGVKLEAN SQKRYLLTGQ ILSDGKVFVH LCNYIEPWEN LSFLQRESLN HHYHLNCGCQ ITTCYAVPCT ISAPNECLWT DWLLERKLYG YQAQHYVCMK HVDGSCSWYQ GRLPLRKEFV DIIQP
Greater than 90% as determined by SDS-PAGE.
317694
100 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol