missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Cytokeratin 17 Protein
A DNA sequence encoding the Bos taurus (Bovine) Cytokeratin 17, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6277-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001098792.1 | |
Cytokeratin 17 Protein | |
Research Use Only | |
KRT17 | |
Recombinant Protein | |
E. coli | |
MTTTIRHFSS GSIKGSSGLA GGSSRSCRVS GSLGGGSCRL GSAGGLGSGL GGSSYSSCYS FGSGGGYGSG GYVSGGYGGG FGGVDGLLVG GEKATMQNLN DRLASYLDKV RALEEANTEL ELKIRDWYQK QAPGPAPDYS SYFKTIEDLR NKIHTATVDN ANLLLQIDNA RLAADDFRTK FETEQALRVS VEADINGLRR VLDELTLARA DLEMQIENLK EELAYLRKNH EEEMKALRGQ VGGEINVEMD AAPGVDLSRI LNEMRDQYEK MAEKNRKDAE DWFFSKTEEL NREVATNSEL VQSGKSEISE LRRTLQALEI ELQSQLSMKA SLEGSLAETE NRYCMQLSQI QGLIGSVEEQ LAQLRCEMEQ QNQEYKILLD VKTRLEQEIA TYRRLLEGED AHLTQYKTKE PVTTRQVRTI VEEVQDGRVI SSREQVHQTS H | |
Greater than 90% as determined by SDS-PAGE. |
281889 | |
1 mg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |