Learn More
Thermo Scientific™ Chiller Plumbing Kits and Accessories
Choose these chiller plumbing kits and accessories, which include flexible tubing, tubing insulation and hose clamps.
Brand: Thermo Scientific™ 0012560
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ Chiller Plumbing Kits and accessories include flexible tubing, tubing insulation, and hose clamps.
Features:
- Include flexible tubing, tubing insulation, and hose clamps
Specifications
Coupling | |
-90°C to +105°C | |
1 each M16 to M16 coupling |
VersaCool | |
Coupler | |
M16 |