missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Chiller Plumbing Kits and Accessories

These Chiller Plumbing Kits includes flexible tubing, tubing insulation and hose clamps.

Brand:  Thermo Scientific™ 0821214

Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 10015381

  • 1053.95 NOK / Each


Estimated Shipment
17-11-2020
Add to basket

Specifications

Specifications

Tubing, Viton
Viton Tubing, 8mm ID custom length
Tubing
ThermoTemp, Polar
8 mm ID per meter
Videos
Documents