Learn More
Retsch™ Reaction Vial Adapter
Retsch™ Reaction Vial Adapter is a CryoMill accessory suitable for use with 2/4/6 reaction vials.
Brand:  Retsch™ 02.706.0303
39415.50 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.48000kg
Product Code. 15382888
  • 41490.00 NOK / Each
Estimated Shipment: 24-07-2024
Description
  • Adapter for use of 2/4/6 reaction vials, 2 mL
Specifications
Specifications
2 mL Vials
CryoMill Reaction Vial