Learn More
Novus Biologicals™ Rat Caspase-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand:  Novus Biologicals™ NBP2-75024
Product Code. 18329530
  • 7400.46 NOK / 1 set
Explore more special offers
Description
Description
The Rat Caspase-3 ELISA Kit (Colorimetric) measures rat Caspase-3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Rat CASP3 in samples. No significant cross-reactivity or interference between Rat CASP3 and analogues was observed.
Biological Fluids, Plasma, Serum
836
Rat
Storage of components varies. See protocol for specific instructions.
0.19 ng/mL
ELISA Kit (Colorimetric)
ELISA
CASP3
1 Kit
Caspase-3