Learn More
Novus Biologicals™ Rat Caspase-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: Novus Biologicals™ NBP2-75024
This item is not returnable.
View return policy
Description
The Rat Caspase-3 ELISA Kit (Colorimetric) measures rat Caspase-3 in serum, plasma and other biological fluids.Specifications
0.31 - 20 ng/mL | |
This kit recognizes Rat CASP3 in samples. No significant cross-reactivity or interference between Rat CASP3 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
836 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.19 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CASP3 | |
1 Kit | |
Caspase-3 |