missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RAIDD Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0004
Additional Details : Weight : 0.01000kg
Description
RAIDD Synthetic Peptide, PEP-0004, from Invitrogen.
This peptide corresponds to 19 amino acids near the center of human RAIDD.PEP-0004 can be used as a blocking peptide with polyclonal antibody PA5-19864.Specifications
8738 | |
purified | |
Human RAIDD | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
P78560 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
CRADD | |
0.02% sodium azide | |
50 μg | |
PBS |