Learn More
Tenak™ Microtiterplates Comfort Rack For Upright Freezers
Rack designed to hold microtiterplates in upright freezers.
Brand: Tenak™ TE35528
Additional Details : Weight : 0.55000kg
Description
- Variety of shelf heights available
- Racks can either be used as one plate per shelf or the plates can be stacked
- Includes an alert preventing the shelves from sliding out
Specifications
Microtiterplate Rack | |
Plates (138) | |
16 mm |
23 x 6 | |
Upright Freezers |