missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ ECO AluRack for Chest Freezers, 100 mm Box Height, with Boxes

ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11564B

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12458150

  • 949.00 NOK / Each


Estimated Shipment: 06-12-2021
Add to basket

Description

Description

  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

Seawater resistant
Chest Frseezers
Boxes (5)
Aluminum
140 x 140 x 530 mm
100 mm (Box)
White cardboard boxes, 9 x 9 cell dividers
Videos
SDS
Documents
Certificates