Learn More
Tenak™ ECO AluRack for Upright Freezers, 100 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11304B
Additional Details : Weight : 2.60000kg
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
3 x 4 | |
Aluminum | |
564 x 139 x 315 mm |
Seawater resistant | |
Boxes (12) | |
Upright Freezers | |
100 mm (Box) |