Learn More
Tenak™ Stainless Steel Feezer Rack for Microtiterplates, with Locking Rod
A range of freezer racks designed for use with microtiterplates.
Brand:  Tenak™ TE27700
Additional Details : Weight : 2.80000kg
 View more versions of this product
Product Code. 12456970
  • 1565.00 NOK / Each
Estimated Shipment: 18-07-2024
Description
  • For use in freezers
  • Includes locking rod
  • Contstructed of stainless steel
Specifications
Specifications
39 mm
Microtitre Plates
Stainless Steel
717 mm
Microtiterplates (18)
Freezer Rack