Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 25 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE15132
Additional Details : Weight : 1.60000kg
 View more versions of this product
Product Code. 12468170
Quantity Price
1 869.00 NOK / Each
10 801.91 NOK / Each
Estimated Shipment: 09-12-2021
Add to basket
Description
Description
  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Specifications
Chest Frseezers
Freezer Rack
135 x 92 x 355 mm
Aluminum
25 mm
Microtiterplates (13)