missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 25 mm Shelf Height

Eco AluRack for upright freezers is designed to hold microtiterplates.

Brand:  Tenak™ TE15416

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 12468210

Quantity Price
1 1472.00 NOK / Each
10 1358.37 NOK / Each
Estimated Shipment: 15-12-2021

Add to basket


Description

Description

  • For use in upright freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications

Specifications

12 x 4
Upright Freezers
Aluminum
360 x 135 x 325 mm
Microtiterplates (48)
25 mm
Videos
SDS
Documents
Certificates