missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac 1-Oleoyl-2-linoleoyl-3-chloropropanediol-d5, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | rac 1-Oleoyl-2-linoleoyl-3-chloropropanediol-d5 |
| Molecular Formula | C39H64D5ClO4 |
| Quantity | 25 mg |
| SMILES | O=C(CCCCCCC/C=C\CCCCCCCC)OC([2H])([2H])C(C([2H])([2H])Cl)([2H])OC(CCCCCCC/C=C\C/C=C\CCCCC)=O |
| Molecular Weight (g/mol) | 642.45 |
| Formula Weight | g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction